Anti-FPR3 Mouse Monoclonal Antibody [clone: 374822]

Supplier: R&D Systems
MAB3896
220020-662EA 634.86 USD
220020-662
Anti-FPR3 Mouse Monoclonal Antibody [clone: 374822]
Antibodies
Type: Primary
Antigen Symbol: FPR3
Antigen Name: formyl peptide receptor 3
Antigen Synonyms: FML2_HUMAN, FMLPY, FPRH1, FPRH2, FPRL2, RMLP-R-I
Clonality: Monoclonal
Clone: 374822
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR