Anti-NRP1 Mouse Monoclonal Antibody [clone: 446915]

Supplier: R&D Systems
MAB38701
220020-650EA 634.86 USD
220020-650
Anti-NRP1 Mouse Monoclonal Antibody [clone: 446915]
Antibodies
Type: Primary
Antigen: NRP1
Clonality: Monoclonal
Clone: 446915
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR