Anti-PDGFRB Mouse Monoclonal Antibody [clone: 61520]

Supplier: R&D Systems
MAB385
220020-643EA 808.4 USD
220020-643
Anti-PDGFRB Mouse Monoclonal Antibody [clone: 61520]
Antibodies
Type: Primary
Antigen: PDGFRB
Clonality: Monoclonal
Clone: 61520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR