Anti-EDN1 Rat Monoclonal Antibody [clone: 3G10]

Supplier: R&D Systems
MAB3440
220020-457EA 692.71 USD
220020-457
Anti-EDN1 Rat Monoclonal Antibody [clone: 3G10]
Antibodies
Direct ELISA

Type: Primary
Antigen: EDN1
Clonality: Monoclonal
Clone: 3G10
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR