Anti-PTGDR2 Mouse Monoclonal Antibody [clone: 301108]

Supplier: R&D Systems
MAB3338
220020-400EA 634.86 USD
220020-400
Anti-PTGDR2 Mouse Monoclonal Antibody [clone: 301108]
Antibodies
Type: Primary
Antigen: PTGDR2
Clonality: Monoclonal
Clone: 301108
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR