Anti-SLC27A1 Mouse Monoclonal Antibody [clone: 308420]

Supplier: R&D Systems
MAB3304
220020-385EA 634.86 USD
220020-385
Anti-SLC27A1 Mouse Monoclonal Antibody [clone: 308420]
Antibodies
Type: Primary
Antigen: SLC27A1
Clonality: Monoclonal
Clone: 308420
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR