Anti-TRAF3 Rat Monoclonal Antibody [clone: 433113]

Supplier: R&D Systems
MAB3278
220020-371EA 504.71 USD
220020-371
Anti-TRAF3 Rat Monoclonal Antibody [clone: 433113]
Antibodies
Type: Primary
Antigen: TRAF3
Clonality: Monoclonal
Clone: 433113
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR