Anti-CA5B Mouse Monoclonal Antibody [clone: 491725]

Supplier: R&D Systems
MAB3176
220020-323EA 577.02 USD
220020-323
Anti-CA5B Mouse Monoclonal Antibody [clone: 491725]
Antibodies
Type: Primary
Antigen: CA5B
Clonality: Monoclonal
Clone: 491725
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR