Anti-AGT Mouse Monoclonal Antibody [clone: 369439]

Supplier: R&D Systems
MAB3156
220020-311EA 620.4 USD
220020-311
Anti-AGT Mouse Monoclonal Antibody [clone: 369439]
Antibodies
Type: Primary
Antigen: AGT
Clonality: Monoclonal
Clone: 369439
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR