Anti-THPO Mouse Monoclonal Antibody [clone: 34863]

Supplier: R&D Systems
MAB2881
220020-156EA 808.4 USD
220020-156
Anti-THPO Mouse Monoclonal Antibody [clone: 34863]
Antibodies
Type: Primary
Antigen: THPO
Clonality: Monoclonal
Clone: 34863
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR