Anti-LPPR4 Mouse Monoclonal Antibody [clone: 368701]

Supplier: R&D Systems
MAB2874
220020-152EA 504.71 USD
220020-152
Anti-LPPR4 Mouse Monoclonal Antibody [clone: 368701]
Antibodies
Type: Primary
Antigen: LPPR4
Clonality: Monoclonal
Clone: 368701
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR