Anti-STC2 Rat Monoclonal Antibody [clone: 348310]

Supplier: R&D Systems
MAB2837
220020-128EA 577.02 USD
220020-128
Anti-STC2 Rat Monoclonal Antibody [clone: 348310]
Antibodies
Type: Primary
Antigen: STC2
Clonality: Monoclonal
Clone: 348310
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR