Anti-ESAM Rat Monoclonal Antibody [clone: 340236]

Supplier: R&D Systems
MAB28271
220020-120EA 620.4 USD
220020-120
Anti-ESAM Rat Monoclonal Antibody [clone: 340236]
Antibodies
Type: Primary
Antigen: ESAM
Clonality: Monoclonal
Clone: 340236
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR