Anti-CCL2 Mouse Monoclonal Antibody [clone: 280707]

Supplier: R&D Systems
MAB2817
220020-113EA 620.4 USD
220020-113
Anti-CCL2 Mouse Monoclonal Antibody [clone: 280707]
Antibodies
Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone: 280707
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR