Anti-MSR1 Mouse Monoclonal Antibody [clone: 351615]

Supplier: R&D Systems
MAB2708
220020-044EA 692.71 USD
220020-044
Anti-MSR1 Mouse Monoclonal Antibody [clone: 351615]
Antibodies
Type: Primary
Antigen: MSR1
Clonality: Monoclonal
Clone: 351615
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR