Anti-LY9 Rat Monoclonal Antibody [clone: 312830]

Supplier: R&D Systems
MAB2555
220019-939EA 620.4 USD
220019-939
Anti-LY9 Rat Monoclonal Antibody [clone: 312830]
Antibodies
Type: Primary
Antigen: LY9
Clonality: Monoclonal
Clone: 312830
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR