Anti-TFPI2 Mouse Monoclonal Antibody [clone: 243220]

Supplier: R&D Systems
MAB2545
220019-930EA 808.4 USD
220019-930
Anti-TFPI2 Mouse Monoclonal Antibody [clone: 243220]
Antibodies
Type: Primary
Antigen: TFPI2
Clonality: Monoclonal
Clone: 243220
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR