Anti-FGF21 Mouse Monoclonal Antibody [clone: 315914]

Supplier: R&D Systems
MAB2537
220019-921EA 620.4 USD
220019-921
Anti-FGF21 Mouse Monoclonal Antibody [clone: 315914]
Antibodies
Type: Primary
Antigen Symbol: FGF21
Antigen Name: fibroblast growth factor 21
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 315914
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR