Anti-MKK3/MKK6 Mouse Monoclonal Antibody [clone: 275922]

Supplier: R&D Systems
MAB2514
220019-906EA 620.4 USD
220019-906
Anti-MKK3/MKK6 Mouse Monoclonal Antibody [clone: 275922]
Antibodies
Type: Primary
Antigen: MKK3/MKK6
Clonality: Monoclonal
Clone: 275922
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR