Anti-LIFR Mouse Monoclonal Antibody [clone: 32953]

Supplier: R&D Systems
MAB249
220019-890EA 837.32 USD
220019-890
Anti-LIFR Mouse Monoclonal Antibody [clone: 32953]
Antibodies
Type: Primary
Antigen: LIFR
Clonality: Monoclonal
Clone: 32953
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR