Anti-RGMA Rat Monoclonal Antibody [clone: 275407]

Supplier: R&D Systems
MAB2458
220019-872EA 634.86 USD
220019-872
Anti-RGMA Rat Monoclonal Antibody [clone: 275407]
Antibodies
Type: Primary
Antigen: RGMA
Clonality: Monoclonal
Clone: 275407
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR