Anti-CHRDL2 Mouse Monoclonal Antibody [clone: 272905]

Supplier: R&D Systems
MAB24481
220019-862EA 605.94 USD
220019-862
Anti-CHRDL2 Mouse Monoclonal Antibody [clone: 272905]
Antibodies
Type: Primary
Antigen: CHRDL2
Clonality: Monoclonal
Clone: 272905
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR