Anti-LILRB4 Mouse Monoclonal Antibody [clone: 293622]

Supplier: R&D Systems
MAB2425
220019-844EA 663.78 USD
220019-844
Anti-LILRB4 Mouse Monoclonal Antibody [clone: 293622]
Antibodies
Type: Primary
Antigen: LILRB4
Clonality: Monoclonal
Clone: 293622
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR