Anti-NCAM1 Mouse Monoclonal Antibody [clone: 301021]

Supplier: R&D Systems
MAB24081
220019-833EA 692.71 USD
220019-833
Anti-NCAM1 Mouse Monoclonal Antibody [clone: 301021]
Antibodies
Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: 301021
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR