Anti-CD69 Mouse Monoclonal Antibody [clone: 298614]

Supplier: R&D Systems
MAB23591
220019-795EA 649.32 USD
220019-795
Anti-CD69 Mouse Monoclonal Antibody [clone: 298614]
Antibodies
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: 298614
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR