Anti-AKT2 Mouse Monoclonal Antibody [clone: 32514]

Supplier: R&D Systems
MAB2315
220019-761EA 577.02 USD
220019-761
Anti-AKT2 Mouse Monoclonal Antibody [clone: 32514]
Antibodies
Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 32514
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR