Anti-WNT8A Rat Monoclonal Antibody [clone: 254126]

Supplier: R&D Systems
MAB2248
220019-710EA 793.94 USD
220019-710
Anti-WNT8A Rat Monoclonal Antibody [clone: 254126]
Antibodies
Type: Primary
Antigen: WNT8A
Clonality: Monoclonal
Clone: 254126
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR