Anti-SEMA6C Mouse Monoclonal Antibody [clone: 254506]

Supplier: R&D Systems
MAB2219
220019-691EA 793.94 USD
220019-691
Anti-SEMA6C Mouse Monoclonal Antibody [clone: 254506]
Antibodies
Type: Primary
Antigen: SEMA6C
Clonality: Monoclonal
Clone: 254506
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR