Anti-ADAMTS5 Mouse Monoclonal Antibody [clone: 36281]

Supplier: R&D Systems
MAB2198
220019-671EA 634.86 USD
220019-671
Anti-ADAMTS5 Mouse Monoclonal Antibody [clone: 36281]
Antibodies
Type: Primary
Antigen: ADAMTS5
Clonality: Monoclonal
Clone: 36281
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR