Anti-TNC Rat Monoclonal Antibody [clone: 578]

Supplier: R&D Systems
MAB2138
220019-626EA 721.63 USD
220019-626
Anti-TNC Rat Monoclonal Antibody [clone: 578]
Antibodies
Type: Primary
Antigen: TNC
Clonality: Monoclonal
Clone: 578
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR