Anti-THY1 Mouse Monoclonal Antibody [clone: Thy-1A1]

Supplier: R&D Systems
MAB2067
220019-562EA 707.17 USD
220019-562
Anti-THY1 Mouse Monoclonal Antibody [clone: Thy-1A1]
Antibodies
Type: Primary
Antigen Symbol: THY1
Antigen Name: Thy-1 cell surface antigen
Antigen Synonyms: CD90, CDw90
Clonality: Monoclonal
Clone: Thy-1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR