Anti-NCR3 Mouse Monoclonal Antibody [clone: 210847]

Supplier: R&D Systems
MAB18491
220019-405EA 837.32 USD
220019-405
Anti-NCR3 Mouse Monoclonal Antibody [clone: 210847]
Antibodies
Agonist Activity

Type: Primary
Antigen: NCR3
Clonality: Monoclonal
Clone: 210847
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR