Anti-NTRK2 Rat Monoclonal Antibody [clone: 225105]

Supplier: R&D Systems
MAB1494
220019-126EA 808.4 USD
220019-126
Anti-NTRK2 Rat Monoclonal Antibody [clone: 225105]
Antibodies
Type: Primary
Antigen: NTRK2
Clonality: Monoclonal
Clone: 225105
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR