Anti-CSTB Rat Monoclonal Antibody [clone: 227818]

Supplier: R&D Systems
MAB1409
220019-059EA 808.4 USD
220019-059
Anti-CSTB Rat Monoclonal Antibody [clone: 227818]
Antibodies
Type: Primary
Antigen: CSTB
Clonality: Monoclonal
Clone: 227818
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR