Anti-TNFSF4 Rat Monoclonal Antibody [clone: 182609]

Supplier: R&D Systems
MAB12362
220018-928EA 808.4 USD
220018-928
Anti-TNFSF4 Rat Monoclonal Antibody [clone: 182609]
Antibodies
Type: Primary
Antigen: TNFSF4
Clonality: Monoclonal
Clone: 182609
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR