Anti-CD40L Rat Monoclonal Antibody [clone: 208109]

Supplier: R&D Systems
MAB1163
220018-856EA 808.4 USD
220018-856
Anti-CD40L Rat Monoclonal Antibody [clone: 208109]
Antibodies
Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: 208109
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR