Anti-PPBP Mouse Monoclonal Antibody [clone: 186701]

Supplier: R&D Systems
MAB1116
220018-808EA 793.94 USD
220018-808
Anti-PPBP Mouse Monoclonal Antibody [clone: 186701]
Antibodies
Type: Primary
Antigen: PPBP
Clonality: Monoclonal
Clone: 186701
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR