Anti-EDA2R Mouse Monoclonal Antibody [clone: 173111]

Supplier: R&D Systems
MAB1093
220018-780EA 822.86 USD
220018-780
Anti-EDA2R Mouse Monoclonal Antibody [clone: 173111]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: EDA2R
Clonality: Monoclonal
Clone: 173111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR