Anti-NEO1 Rat Monoclonal Antibody [clone: 221519]

Supplier: R&D Systems
MAB1079
220018-768EA 808.4 USD
220018-768
Anti-NEO1 Rat Monoclonal Antibody [clone: 221519]
Antibodies
Type: Primary
Antigen Symbol: NEO1
Antigen Name: neogenin
Antigen Synonyms: 2610028H22Rik, AI327052, D930014N22Rik, Igdcc2
Clonality: Monoclonal
Clone: 221519
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR