Anti-BMP8A Mouse Monoclonal Antibody [clone: 158708]

Supplier: R&D Systems
MAB1073
220018-762EA 822.86 USD
220018-762
Anti-BMP8A Mouse Monoclonal Antibody [clone: 158708]
Antibodies
Type: Primary
Antigen: BMP8A
Clonality: Monoclonal
Clone: 158708
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR