Anti-EREG Rat Monoclonal Antibody [clone: 189611]

Supplier: R&D Systems
MAB1068
220018-755EA 793.94 USD
220018-755
Anti-EREG Rat Monoclonal Antibody [clone: 189611]
Antibodies
Type: Primary
Antigen: EREG
Clonality: Monoclonal
Clone: 189611
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR