Anti-IL6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1936]

Supplier: R&D Systems
IC206P
220018-290EA 605.94 USD
220018-290
Anti-IL6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1936]
Antibodies
Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone: 1936
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR