Anti-IL1B Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 8516]

Supplier: R&D Systems
IC201C
220018-271EA 663.78 USD
220018-271
Anti-IL1B Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 8516]
Antibodies
Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone: 8516
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR