Anti-IL1A Mouse Monoclonal Antibody (Fluorescein) [clone: 4414]

Supplier: R&D Systems
IC200F
220018-266EA 475.78 USD
220018-266
Anti-IL1A Mouse Monoclonal Antibody (Fluorescein) [clone: 4414]
Antibodies
Type: Primary
Antigen: IL1A
Clonality: Monoclonal
Clone: 4414
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR