Anti-ADAM10 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1633]

Supplier: R&D Systems
IC1427P
220018-235EA 562.55 USD
220018-235
Anti-ADAM10 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1633]
Antibodies
Type: Primary
Antigen: ADAM10
Clonality: Monoclonal
Clone: 1633
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR