Mouse IgG2a Isotype Control

Supplier: R&D Systems
IC003A
220018-149EA 345.63 USD
220018-149
Mouse IgG2a Isotype Control
Antibodies
Selected for use as mouse IgG2a isotype control for flow cytometry applications.

Type: Control
Antigen: IgG2a
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR