Anti-CKM Goat Polyclonal Antibody

Supplier: R&D Systems
G-113-C
220018-094EA 577.02 USD
220018-094
Anti-CKM Goat Polyclonal Antibody
Antibodies
Hidden

Type: Primary
Antigen: CKM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR