Anti-IgM µ Goat Polyclonal Antibody

Supplier: R&D Systems
G-105-C
220018-091EA 678.25 USD
220018-091
Anti-IgM µ Goat Polyclonal Antibody
Antibodies
Hidden

Type: Primary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR