Anti-ACE Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 171417]

Supplier: R&D Systems
FAB929P
220018-021EA 519.17 USD
220018-021
Anti-ACE Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 171417]
Antibodies
Type: Primary
Antigen: ACE
Clonality: Monoclonal
Clone: 171417
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR