Anti-TNFSF18 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 109101]

Supplier: R&D Systems
FAB6941A
220017-970EA 678.25 USD
220017-970
Anti-TNFSF18 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 109101]
Antibodies
Type: Primary
Antigen: TNFSF18
Clonality: Monoclonal
Clone: 109101
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR